Lead(II) chloride for perovskite precursor, purified by sublimation

Supplier: TCI America
Danger

L0291-1G L0291-5G
TCL0291-1GEA 70 USD
TCL0291-1G TCL0291-5G
Lead(II) chloride for perovskite precursor, purified by sublimation
Lead(II) chloride
Formula: Cl₂Pb
Boiling Pt: 951 °C
Melting Pt: 501 °C
MDL Number: MFCD00011157
CAS Number: 7758-95-4
EINECS: 231-845-5
UN: 2291
ADR: 6.1,III
Merck Index: 14,05404

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR