2-Iodo-5-nitrotoluene ≥96.0% (by GC)
Supplier: TCI America
|
Warning
|
I0992-25G
I0992-5G
I0992-1G
TCI0992-25GEA
619
USD
TCI0992-25G
TCI0992-5G
TCI0992-1G
2-Iodo-5-nitrotoluene ≥96.0% (by GC)
2-Iodo-5-nitrotoluene
Formula:
C₇H₆INO₂ |
MDL Number:
MFCD00051517 CAS Number: 5326-38-5 EINECS: 226-204-1 |
Specification Test Results
Appearance | Light yellow to Brown powder to crystal |
Purity (GC) | min. 96.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...