2-(Dimethylamino)benzoic acid ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 2-Dimethylaminobenzoic acid

D5463-25G D5463-5G
TCD5463-25GEA 1174 USD
TCD5463-25G TCD5463-5G
2-(Dimethylamino)benzoic acid ≥98.0% (by HPLC)
2-(Dimethylamino)benzoic acid
Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00016496
CAS Number: 610-16-2
EINECS: 210-209-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %
Melting point 68.0 to 72.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR