9-Vinylanthracene ≥98.0% (by GC)

Supplier: TCI America

V0130-1G V0130-5G
TCV0130-1GEA 64 USD
TCV0130-1G TCV0130-5G
9-Vinylanthracene ≥98.0% (by GC)
9-Vinylanthracene
Formula: C₁₆H₁₂
Boiling Pt: 145 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00003576
CAS Number: 2444-68-0
EINECS: 219-486-2

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR