n-Pentane ≥97.0% (by GC)

Supplier: TCI America
Danger

P2621-100ML P2621-500ML
TCP2621-100MLEA 18 USD
TCP2621-100ML TCP2621-500ML
n-Pentane ≥97.0% (by GC)
n-Pentane
Formula: H₃C(CH₂)₃CH₃
Boiling Pt: 36 °C
Flash Pt: −49 °C
MDL Number: MFCD00009498
CAS Number: 109-66-0
EINECS: 203-692-4
UN: 1265
ADR: 3,II
Merck Index: 14,07116

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR