Trimethylamine ∼13% ∼2 mol/L in acetonitrile

Supplier: TCI America
Danger

T3567-500ML T3567-100ML
TCT3567-500MLEA 124 USD
TCT3567-500ML TCT3567-100ML
Trimethylamine ∼13% ∼2 mol/L in acetonitrile
Trimethylamine
Formula: C₃H₉N
Storage Temperature: Refrigerator
MDL Number: MFCD00008327
CAS Number: 75-50-3
EINECS: 200-875-0
UN: 3286
ADR: 3,II
Merck Index: 14,09710

Order Now

Specification Test Results

Appearance Colourless to Almost Colourless clear liquid
Concentration (Neutralization titration) 13.0 to 15.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR