2,6-Dichloro-4-pyridineboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2,6-Dichloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2,6-Dichloropyridine-4-boronic acid Pinacol ester, 2,6-Dichloropyridine-4-boronic acid, pinacol ester, 2-(2,6-Dichloro-4-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

D5574-1G D5574-5G
TCD5574-1GEA 57 USD
TCD5574-1G TCD5574-5G
2,6-Dichloro-4-pyridineboronic acid pinacol ester ≥98.0% (by GC)
2,6-Dichloro-4-pyridineboronic acid pinacol ester
Formula: C₁₁H₁₄BCl₂NO₂
MW: 273.95 g/mol
MDL Number: MFCD09746195
CAS Number: 408492-27-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 96.0 %
Melting point 116.0 to 120.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR