2-tert-Butoxypyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-(tert-Butoxy)pyridine

B5575-1G B5575-5G
TCB5575-1GEA 102 USD
TCB5575-1G TCB5575-5G
2-tert-Butoxypyridine ≥98.0% (by GC)
2-tert-Butoxypyridine
Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 88 °C
MDL Number: MFCD00234567
CAS Number: 83766-88-5

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR