Trihexyl O-Butyrylcitrate ≥92.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Butyryl trihexyl Citrate, Butyrylcitric acid trihexyl ester, Trihexyl 2-(Butyryloxy)propane-1,2,3-tricarboxylate, 2-(Butyryloxy)propane-1,2,3-tricarboxylic acid trihexyl ester

B6040-25G B6040-500G
TCB6040-25GEA 30 USD
TCB6040-25G TCB6040-500G
Trihexyl O-Butyrylcitrate ≥92.0% (by GC)
Trihexyl O-Butyrylcitrate
Formula: C₂₈H₅₀O₈
MW: 514.70 g/mol
Flash Pt: 234 °C
MDL Number: MFCD00192535
CAS Number: 82469-79-2
EINECS: 413-890-4
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 92.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR