4-(Bromomethyl)-4'-methyl-2,2'-bipyridyl ≥97.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 4-(Bromomethyl)-4'-methyl-2,2'-bipyridine

B3986-1G B3986-200MG
TCB3986-1GEA 524 USD
TCB3986-1G TCB3986-200MG
4-(Bromomethyl)-4'-methyl-2,2'-bipyridyl ≥97.0% (by GC)
4-(Bromomethyl)-4'-methyl-2,2'-bipyridyl
Formula: C₁₂H₁₁BrN₂
MW: 263.14 g/mol
MDL Number: MFCD08275265
CAS Number: 81998-05-2
UN: 3261
ADR: 8,III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystaline
Purity (GC) min. 97.0 %
Melting point 74.0 to 78.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR