Bis[2-[(oxo)diphenylphosphino]phenyl] ether ≥99.0% (by HPLC), purified by sublimation

Supplier: TCI America

Synonyms: (Oxybis(2,1-phenylene))bis(diphenylphosphine oxide), DPEPO

B5448-1G B5448-200MG
TCB5448-1GEA 202 USD
TCB5448-1G TCB5448-200MG
Bis[2-[(oxo)diphenylphosphino]phenyl] ether ≥99.0% (by HPLC), purified by sublimation
Bis[2-[(oxo)diphenylphosphino]phenyl] ether
Formula: C₃₆H₂₈O₃P₂
MW: 570.56 g/mol
Melting Pt: 280 °C
MDL Number: MFCD28968111
CAS Number: 808142-23-6

Order Now

Specification Test Results

Purity (HPLC) min. 99.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR