4-Bromo-2,6-diisopropylaniline ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

B4113-1G B4113-5G
TCB4113-1GEA 40 USD
TCB4113-1G TCB4113-5G
4-Bromo-2,6-diisopropylaniline ≥98.0% (by titrimetric analysis)
4-Bromo-2,6-diisopropylaniline
Formula: C₁₂H₁₈BrN
Boiling Pt: 110 °C
MDL Number: MFCD09743895
CAS Number: 80058-84-0

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 95.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR