1,3-Bis(trifluoromethyl)cyclohexane cis- and trans-mixture ≥98.0% (by GC)

Supplier: TCI America
Warning

B5645-1G B5645-5G
TCB5645-1GEA 60 USD
TCB5645-1G TCB5645-5G
1,3-Bis(trifluoromethyl)cyclohexane cis- and trans-mixture ≥98.0% (by GC)
1,3-Bis(trifluoromethyl)cyclohexane
Formula: C₈H₁₀F₆
MW: 220.16 g/mol
Boiling Pt: 135 °C
CAS Number: 79850-12-7
UN: 1993
ADR: 3 III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR