2-Chloro-4,6-di(naphthalen-1-yl)-1,3,5-triazine ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C3525-1G
C3525-200MG
TCC3525-1GEA
257
USD
TCC3525-1G
TCC3525-200MG
2-Chloro-4,6-di(naphthalen-1-yl)-1,3,5-triazine ≥98.0% (by GC)
2-Chloro-4,6-di(naphthalen-1-yl)-1,3,5-triazine
Formula:
C₂₃H₁₄ClN₃ |
CAS Number:
78941-32-9 |
Specification Test Results
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 148.0 to 152.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...