Cyanomethyl Methyl(phenyl)carbamodithioate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: Methyl(phenyl)carbamodithioic acid Cyanomethyl ester

C3534-1G C3534-5G
TCC3534-1GEA 157 USD
TCC3534-1G TCC3534-5G
Cyanomethyl Methyl(phenyl)carbamodithioate ≥98.0% (by HPLC, total nitrogen)
Cyanomethyl Methyl(phenyl)carbamodithioate
Formula: C₁₀H₁₀N₂S₂
MDL Number: MFCD16038113
CAS Number: 76926-16-4

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR