1,4-Diethynyl-2,5-dimethylbenzene ≥98.0% (by GC)

Supplier: TCI America

D5314-1G D5314-200MG
TCD5314-1GEA 552 USD
TCD5314-1G TCD5314-200MG
1,4-Diethynyl-2,5-dimethylbenzene ≥98.0% (by GC)
1,4-Diethynyl-2,5-dimethylbenzene
Formula: C₁₂H₁₀
MW: 154.21 g/mol
CAS Number: 75867-45-7

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 110.0 to 114.0 deg-C
Solubility in Toluene almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR