9-(2-Bromoethyl)carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5182-1G B5182-5G
TCB5182-1GEA 106 USD
TCB5182-1G TCB5182-5G
9-(2-Bromoethyl)carbazole ≥98.0% (by GC)
9-(2-Bromoethyl)carbazole
Formula: C₁₄H₁₂BrN
MW: 274.16 g/mol
MDL Number: MFCD11127041
CAS Number: 75620-93-8

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR