2-Acetamido-3-methylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

A3065-1G A3065-5G
TCA3065-1GEA 61 USD
TCA3065-1G TCA3065-5G
2-Acetamido-3-methylpyridine ≥98.0% (by GC)
2-Acetamido-3-methylpyridine
Formula: C₈H₁₀N₂O
Boiling Pt: 158 °C
MDL Number: MFCD00495347
CAS Number: 7463-30-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 90.0 to 94.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR