1,4-Diethynyl-2,5-dimethoxybenzene ≥98.0% (by GC)

Supplier: TCI America

D5315-1G D5315-200MG
TCD5315-1GEA 195 USD
TCD5315-1G TCD5315-200MG
1,4-Diethynyl-2,5-dimethoxybenzene ≥98.0% (by GC)
1,4-Diethynyl-2,5-dimethoxybenzene
Formula: C₁₂H₁₀O₂
MW: 186.21 g/mol
Melting Pt: 166 °C
MDL Number: MFCD00846857
CAS Number: 74029-40-6

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR