2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(2-phenylpropan-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol ≥98.0% (by HPLC)

Supplier: TCI America

B5976-25G B5976-5G
TCB5976-25GEA 116 USD
TCB5976-25G TCB5976-5G
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(2-phenylpropan-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol ≥98.0% (by HPLC)
2-(2H-Benzo[d][1,2,3]triazol-2-yl)-6-(2-phenylpropan-2-yl)-4-(2,4,4-trimethylpentan-2-yl)phenol
Formula: C₂₉H₃₅N₃O
MDL Number: MFCD02100741
CAS Number: 73936-91-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 110.0 to 114.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR