11-(Bromomethyl)tricosane ≥95.0% (by GC)

Supplier: TCI America
Warning

B5691-25ML B5691-5ML
TCB5691-25MLEA 687 USD
TCB5691-25ML TCB5691-5ML
11-(Bromomethyl)tricosane ≥95.0% (by GC)
11-(Bromomethyl)tricosane
Formula: C₂₄H₄₉Br
MW: 417.56 g/mol
Storage Temperature: Refrigerator
CAS Number: 732276-63-0

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR