(S)-(+)-2-Chloro-1-phenylethanol ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (S)-2-Chloro-1-phenylethanol, (S)-(+)-alpha-(Chloromethyl)benzyl alcohol

C3562-5G C3562-25G
TCC3562-5GEA 85 USD
TCC3562-5G TCC3562-25G
(S)-(+)-2-Chloro-1-phenylethanol ≥98.0% (by GC)
(S)-(+)-2-Chloro-1-phenylethanol
Formula: C₈H₉ClO
Boiling Pt: 114 °C
MDL Number: MFCD00077692
CAS Number: 70111-05-6

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Optical purity (GC) min. 98.0 ee%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR