5-Bromo-6-hydroxy-1,3-benzodioxole ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-4,5-methylenedioxyphenol, 6-Bromobenzo[d][1,3]dioxol-5-ol

B4526-1G B4526-5G
TCB4526-1GEA 202 USD
TCB4526-1G TCB4526-5G
5-Bromo-6-hydroxy-1,3-benzodioxole ≥98.0% (by HPLC)
5-Bromo-6-hydroxy-1,3-benzodioxole
Formula: C₇H₅BrO₃
MW: 217.02 g/mol
MDL Number: MFCD21364431
CAS Number: 6941-70-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 77.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR