4-Aminocyclohexanone hydrochloride ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Warning
|
A3070-1G
A3070-5G
TCA3070-1GEA
127
USD
TCA3070-1G
TCA3070-5G
4-Aminocyclohexanone hydrochloride ≥98.0% (by titrimetric analysis)
4-Aminocyclohexanone hydrochloride
Formula:
C₆H₁₁NO·HCl |
MDL Number:
MFCD06659765 CAS Number: 675112-40-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Nonaqueous Titration) | min. 98.0 % |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...