tert-Butyl 4-Formylbenzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: tert-Butyl 4-formylbenzoate, 4-Formylbenzoic acid tert-Butyl ester, 4-(tert-Butoxycarbonyl)benzaldehyde

B5414-1G B5414-5G
TCB5414-1GEA 216.15 USD
TCB5414-1G TCB5414-5G
tert-Butyl 4-Formylbenzoate ≥98.0% (by GC)
tert-Butyl 4-Formylbenzoate
Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
MDL Number: MFCD01111994
CAS Number: 65874-27-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR