N-(4-Biphenylyl)-2-naphthylamine ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Warning

Synonyms: N-([1,1'-Biphenyl]-4-yl)naphthalen-2-amine

B6030-1G B6030-5G
TCB6030-1GEA 101 USD
TCB6030-1G TCB6030-5G
N-(4-Biphenylyl)-2-naphthylamine ≥98.0% (by HPLC, total nitrogen)
N-(4-Biphenylyl)-2-naphthylamine
Formula: C₂₂H₁₇N
MW: 295.39 g/mol
MDL Number: MFCD08097463
CAS Number: 6336-92-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 144.0 to 148.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR