Calcifediol monohydrate ≥98.0% (by HPLC)
Supplier: TCI America
|
Danger
|
C3537-10MG
C3537-50MG
TCC3537-10MGEA
198
USD
TCC3537-10MG
TCC3537-50MG
Calcifediol monohydrate ≥98.0% (by HPLC)
Calcifediol monohydrate
Formula:
C₂₇H₄₄O₂·H₂O |
MDL Number:
MFCD08600001 CAS Number: 63283-36-3 UN: 2811 ADR: 6.1,II |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Carbon) | 75.50 to 80.00 % |
Water | 3.0 to 5.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...