3-Bromo-9-(2-ethylhexyl)-9H-carbazole ≥98.0% (by GC)

Supplier: TCI America
Warning

B5612-1G B5612-5G
TCB5612-1GEA 117 USD
TCB5612-1G TCB5612-5G
3-Bromo-9-(2-ethylhexyl)-9H-carbazole ≥98.0% (by GC)
3-Bromo-9-(2-ethylhexyl)-9H-carbazole
Formula: C₂₀H₂₄BrN
MDL Number: MFCD18252906
CAS Number: 628336-85-6

Order Now

Specification Test Results

Appearance Colorless to Brown clear liquid
Purity (GC) min. 98.000 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR