2-[9,10-Di(naphthalen-2-yl)anthracen-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC)

Supplier: TCI America

D5307-1G D5307-200MG
TCD5307-1GEA 408 USD
TCD5307-1G TCD5307-200MG
2-[9,10-Di(naphthalen-2-yl)anthracen-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC)
2-[9,10-Di(naphthalen-2-yl)anthracen-2-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₄₀H₃₃BO₂
MDL Number: MFCD26401780
CAS Number: 624744-67-8

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR