2-Bromodiphenylamine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5744-1G B5744-5G
TCB5744-1GEA 122 USD
TCB5744-1G TCB5744-5G
2-Bromodiphenylamine ≥98.0% (by GC)
2-Bromodiphenylamine
Formula: C₁₂H₁₀BrN
Boiling Pt: 141 °C
MDL Number: MFCD01851817
CAS Number: 61613-22-7

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR