2-(3-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 3-Bromophenylboronic acid Pinacol ester, 1-Bromo-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

B6024-1G B6024-5G
TCB6024-1GEA 40 USD
TCB6024-1G TCB6024-5G
2-(3-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by GC, titration analysis)
2-(3-Bromophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formula: C₁₂H₁₆BBrO₂
MDL Number: MFCD12545926
CAS Number: 594823-67-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR