(2-Acetoxyethoxy)methyl Acetate ≥96.0% (by GC)

Supplier: TCI America

A3163-1G A3163-5G
TCA3163-1GEA 30 USD
TCA3163-1G TCA3163-5G
(2-Acetoxyethoxy)methyl Acetate ≥96.0% (by GC)
(2-Acetoxyethoxy)methyl Acetate
Formula: C₇H₁₂O₅
Boiling Pt: 116 °C
MDL Number: MFCD09031327
CAS Number: 59278-00-1
EINECS: 261-686-7

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %
Acid Value max. 3.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR