2-Decyl-1-tetradecanol ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Decyltetradecan-1-ol

D5283-25ML D5283-500ML
TCD5283-25MLEA 38 USD
TCD5283-25ML TCD5283-500ML
2-Decyl-1-tetradecanol ≥97.0% (by GC)
2-Decyl-1-tetradecanol
Formula: C₂₄H₅₀O
MW: 354.66 g/mol
Boiling Pt: 177 °C
Flash Pt: 209 °C
MDL Number: MFCD00674096
CAS Number: 58670-89-6
EINECS: 261-385-0

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR