tert-Butyl 1-(Tosyloxy)-3,6,9,12-tetraoxapentadecan-15-oate ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
|
Warning
|
B5902-1G
B5902-5G
TCB5902-1GEA
119
USD
TCB5902-1G
TCB5902-5G
tert-Butyl 1-(Tosyloxy)-3,6,9,12-tetraoxapentadecan-15-oate ≥98.0% (by HPLC, titration analysis)
tert-Butyl 1-(Tosyloxy)-3,6,9,12-tetraoxapentadecan-15-oate
Formula:
C₂₂H₃₆O₉S |
MDL Number:
MFCD22574810 CAS Number: 581065-94-3 |
Specification Test Results
Appearance | Colorless to Light yellow clear liquid |
Purity (HPLC) | min. 98.0 area% |
Purity (Ester Value) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...