2-(2-Amino-4-nitroanilino)ethanol ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Amino-4-nitro-N-(2-hydroxyethyl)aniline, HC Yellow No. 5

A3332-25G A3332-5G
TCA3332-25GEA 90 USD
TCA3332-25G TCA3332-5G
2-(2-Amino-4-nitroanilino)ethanol ≥98.0% (by GC, titration analysis)
2-(2-Amino-4-nitroanilino)ethanol
Formula: C₈H₁₁N₃O₃
MDL Number: MFCD00156329
CAS Number: 56932-44-6
EINECS: 260-450-0

Order Now

Specification Test Results

Appearance White to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 133.0 to 137.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR