2,5-Bis(4-bromophenyl)-3,4-diphenylcyclopenta-2,4-dienone ≥98.0% (by HPLC)

Supplier: TCI America

B5987-1G B5987-5G
TCB5987-1GEA 82 USD
TCB5987-1G TCB5987-5G
2,5-Bis(4-bromophenyl)-3,4-diphenylcyclopenta-2,4-dienone ≥98.0% (by HPLC)
2,5-Bis(4-bromophenyl)-3,4-diphenylcyclopenta-2,4-dienone
Formula: C₂₉H₁₈Br₂O
MW: 542.27 g/mol
CAS Number: 54523-24-9

Order Now

Specification Test Results

Appearance Red to Dark blue to Black powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 247.0 to 251.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR