4,7-Dibromo-2,1,3-benzoxadiazole ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4,7-Dibromobenzo[c][1,2,5]oxadiazole, 4,7-Dibromobenzofurazan
D4614-1G
D4614-200MG
TCD4614-1GEA
259
USD
TCD4614-1G
TCD4614-200MG
4,7-Dibromo-2,1,3-benzoxadiazole ≥98.0% (by GC)
4,7-Dibromo-2,1,3-benzoxadiazole
Formula:
C₆H₂Br₂N₂O MW: 277.90 g/mol |
MDL Number:
MFCD02114623 CAS Number: 54286-63-4 |
Specification Test Results
Appearance | White to Yellow to Orange powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 111.0 to 115.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...