2-(4-Bromophenyl)-4-phenylquinazoline ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

B5829-1G B5829-200MG
TCB5829-1GEA 323 USD
TCB5829-1G TCB5829-200MG
2-(4-Bromophenyl)-4-phenylquinazoline ≥98.0% (by HPLC, total nitrogen)
2-(4-Bromophenyl)-4-phenylquinazoline
Formula: C₂₀H₁₃BrN₂
MDL Number: MFCD27979424
CAS Number: 540466-42-0

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 194.0 to 198.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR