2,6-Dibromo-9,10-diphenylanthracene ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
D4986-200MG
D4986-1G
TCD4986-200MGEA
133
USD
TCD4986-200MG
TCD4986-1G
2,6-Dibromo-9,10-diphenylanthracene ≥98.0% (by HPLC, titration analysis)
2,6-Dibromo-9,10-diphenylanthracene
Formula:
C₂₆H₁₆Br₂ |
CAS Number:
528609-98-5 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Argentmetric Titration) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...