Bis[2-[2-(2-chloroethoxy)ethoxy]ethyl]ether ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 1,17-Dichloro-3,6,9,12,15-pentaoxaheptadecane

B5839-1G B5839-5G
TCB5839-1GEA 90 USD
TCB5839-1G TCB5839-5G
Bis[2-[2-(2-chloroethoxy)ethoxy]ethyl]ether ≥97.0% (by GC)
Bis[2-[2-(2-chloroethoxy)ethoxy]ethyl]ether
Formula: C₁₂H₂₄Cl₂O₅
MW: 319.22 g/mol
Boiling Pt: 162 °C
Melting Pt: -12 °C
MDL Number: MFCD00068710
CAS Number: 52559-90-7

Order Now

Specification Test Results

Appearance White to Yellow to Green clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR