1,6-Bis(triethoxysilyl)hexane ≥96.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 4,4,11,11-Tetraethoxy-3,12-dioxa-4,11-disilatetradecane

B6035-1G B6035-5G
TCB6035-1GEA 56 USD
TCB6035-1G TCB6035-5G
1,6-Bis(triethoxysilyl)hexane ≥96.0% (by GC)
1,6-Bis(triethoxysilyl)hexane
Formula: C₁₈H₄₂O₆Si₂
MW: 410.70 g/mol
Boiling Pt: 115 °C
MDL Number: MFCD21596675
CAS Number: 52034-16-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR