Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Dimethyl (3,3-difluoro-2-oxoheptyl)phosphonate, (3,3-Difluoro-2-oxoheptyl)phosphonic acid Dimethyl ester

D5176-1G D5176-200MG
TCD5176-1GEA 235 USD
TCD5176-1G TCD5176-200MG
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate ≥98.0% (by GC)
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate
Formula: C₉H₁₇F₂O₄P
MW: 258.20 g/mol
Boiling Pt: 110 °C
MDL Number: MFCD11707108
CAS Number: 50889-46-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR