(E)-Cinnamyl Benzoate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Benzoic acid (E)-Cinnamyl ester, Benzoic acid (E)-3-Phenyl-2-propenyl ester, (E)-3-Phenyl-2-propenyl Benzoate

C3650-100G C3650-25G
TCC3650-100GEA 224 USD
TCC3650-100G TCC3650-25G
(E)-Cinnamyl Benzoate ≥98.0% (by GC)
(E)-Cinnamyl Benzoate
Formula: C₁₆H₁₄O₂
MW: 238.29 g/mol
Boiling Pt: 335 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00475021
CAS Number: 50555-04-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 37.0 to 41.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR