4-Bromo-4'-phenyltriphenylamine ≥98.0% (by GC)

Supplier: TCI America

Synonyms: N-(4-Bromophenyl)-N-phenyl-[1,1'-biphenyl]-4-amine

B4812-1G B4812-5G
TCB4812-1GEA 96 USD
TCB4812-1G TCB4812-5G
4-Bromo-4'-phenyltriphenylamine ≥98.0% (by GC)
4-Bromo-4'-phenyltriphenylamine
Formula: C₂₄H₁₈BrN
MW: 400.32 g/mol
MDL Number: MFCD24387484
CAS Number: 503299-24-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR