Butyltriethoxysilane ≥94.0% (by GC)

Supplier: TCI America
Warning

B5858-25ML B5858-5ML
TCB5858-25MLEA 114 USD
TCB5858-25ML TCB5858-5ML
Butyltriethoxysilane ≥94.0% (by GC)
Butyltriethoxysilane
Formula: C₁₀H₂₄O₃Si
Boiling Pt: 193 °C
MDL Number: MFCD00138096
CAS Number: 4781-99-1

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR