1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)

Supplier: TCI America

Synonyms: [(2,5-Dibromo-1,4-phenylene)bis(ethyne-2,1-diyl)]bis(trimethylsilane), [2-[2,5-Dibromo-4-[2-(trimethylsilyl)ethynyl]phenyl]ethynyl]trimethylsilane

D5316-1G D5316-5G
TCD5316-1GEA 165 USD
TCD5316-1G TCD5316-5G
1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene ≥98.0% (by GC)
1,4-Dibromo-2,5-bis[2-(trimethylsilyl)ethynyl]benzene
Formula: C₁₆H₂₀Br₂Si₂
MW: 428.31 g/mol
MDL Number: MFCD11521283
CAS Number: 478190-79-3

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 119.0 to 123.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR