Dehydrocostus Lactone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: (3aS,6aR,9aR,9bS)-3,6,9-Trimethylenedecahydroazuleno[4,5-b]furan-2(3H)-one, (3aS,6aR,9aR,9bS)-3,6,9-Trimethylenedecahydroazuleno[4,5-b]furan-2(9bH)-one

D5366-10MG D5366-50MG
TCD5366-10MGEA 74 USD
TCD5366-10MG TCD5366-50MG
Dehydrocostus Lactone ≥98.0% (by GC)
Dehydrocostus Lactone
Formula: C₁₅H₁₈O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00210277
CAS Number: 477-43-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR