N-(2-Acryloyloxyethyl)-N-benzyl-N,N-dimethylammonium chloride ∼75% in water stabilized

Supplier: TCI America


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A3310-25G A3310-500G
TCA3310-25GEA 30 USD
TCA3310-25G TCA3310-500G
N-(2-Acryloyloxyethyl)-N-benzyl-N,N-dimethylammonium chloride ∼75% in water stabilized
N-(2-Acryloyloxyethyl)-N-benzyl-N,N-dimethylammonium chloride
Formula: C₁₄H₂₀ClNO₂
MW: 269.77 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD04038840
CAS Number: 46830-22-2
EINECS: 256-283-8

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (HPLC) min. 90.0 area%
Purity (Argentmetric Titration) 74.0 to 77.0 %
Water 23.0 to 26.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR