Chlorotriethoxysilane ≥95.0% (by GC)

Supplier: TCI America
Danger

C3354-25G C3354-5G
TCC3354-25GEA 167 USD
TCC3354-25G TCC3354-5G
Chlorotriethoxysilane ≥95.0% (by GC)
Chlorotriethoxysilane
Formula: C₆H₁₅ClO₃Si
Boiling Pt: 157 °C
Flash Pt: 47 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00015124
CAS Number: 4667-99-6
EINECS: 225-112-9
UN: 2986
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR