Dimethyl 3,5-Pyridinedicarboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Dimethyl pyridine-3,5-dicarboxylate, 3,5-Pyridinedicarboxylic acid Dimethyl ester

D5364-25G D5364-5G
TCD5364-25GEA 295 USD
TCD5364-25G TCD5364-5G
Dimethyl 3,5-Pyridinedicarboxylate ≥98.0% (by GC, titration analysis)
Dimethyl 3,5-Pyridinedicarboxylate
Formula: C₉H₉NO₄
MW: 195.17 g/mol
Boiling Pt: 132 °C
MDL Number: MFCD00518827
CAS Number: 4591-55-3
EINECS: 224-981-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 83.0 to 87.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR